Assay Details
Gene Symbol: Brf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028193.3 | NP_082469.2 | 13 - 14 | 1718 | 70 | |
GenBank mRNA | BC037224.1 | - | 13 - 14 | 1501 | 3257364 | 70 |
BC058112.1 | - | 3 - 4 | 314 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
