Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009396.2 | NP_033422.2 | 5 - 6 | 1240 | 71 | |
GenBank mRNA | AK154405.1 | - | 5 - 6 | 1240 | 71 | |
AK170719.1 | - | 5 - 6 | 1240 | 71 | ||
AK172653.1 | - | 5 - 6 | 1240 | 71 | ||
BC138106.1 | - | 4 - 5 | 1149 | 8861125 | 71 | |
BC138107.1 | - | 4 - 5 | 1149 | 8861126 | 71 | |
L24118.1 | - | 4 - 5 | 1184 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
