Assay Details
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252533.1 | NP_001239462.1 | 9 - 10 | 1058 | 117 | |
NM_023116.4 | NP_075605.1 | 10 - 11 | 1189 | 117 | ||
NR_045533.1 | 9 - 10 | 1133 | 117 | |||
GenBank mRNA | AK034054.1 | - | 10 - 11 | 1190 | 117 | |
AM259383.2 | - | 9 - 10 | 1058 | 117 | ||
BC109156.1 | - | 10 - 11 | 1121 | 40047524 | 117 | |
BC109157.1 | - | 10 - 11 | 1121 | 40047531 | 117 | |
KJ789960.1 | - | 9 - 10 | 777 | 117 | ||
L20343.1 | - | 9 - 10 | 968 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
