Assay Details
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008074.2 | NP_032100.2 | 5 - 6 | 675 | 95 | |
GenBank mRNA | X59300.1 | - | 5 - 6 | 605 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
