Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028119.5 | NP_082395.2 | 2 - 3 | 513 | 85 | |
GenBank mRNA | AK158770.1 | - | 2 - 3 | 393 | 85 | |
BC099410.1 | - | 2 - 3 | 280 | 30933658 | 85 | |
CK627126.1 | - | 2 - 3 | 513 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
