Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028119.5 | NP_082395.2 | 3 - 4 | 707 | 69 | |
GenBank mRNA | AY027937.1 | - | 3 - 4 | 625 | 69 | |
BC028955.1 | - | 2 - 3 | 891 | 69 | ||
BC080821.1 | - | 2 - 3 | 1043 | 69 | ||
BU515611.1 | - | 1 - 2 | 213 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
