Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028119.5 | NP_082395.2 | 5 - 6 | 950 | 78 | |
GenBank mRNA | AY027937.1 | - | 5 - 6 | 868 | 78 | |
BC028955.1 | - | 4 - 5 | 1134 | 78 | ||
BC058589.1 | - | 2 - 3 | 1322 | 78 | ||
BC080821.1 | - | 4 - 5 | 1286 | 78 | ||
BU515611.1 | - | 3 - 4 | 456 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
