Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028119.5 | NP_082395.2 | 6 - 7 | 1134 | 99 | |
GenBank mRNA | AY027937.1 | - | 6 - 7 | 1052 | 99 | |
BC028955.1 | - | 5 - 6 | 1318 | 99 | ||
BC058589.1 | - | 3 - 4 | 1506 | 99 | ||
BC080821.1 | - | 5 - 6 | 1470 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
