Assay Details
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164072.1 | NP_001157544.1 | 1 - 2 | 113 | 70 | |
GenBank mRNA | AK031559.1 | - | 1 - 2 | 75 | 70 | |
AK038910.1 | - | 1 - 2 | 48 | 70 | ||
AK088817.1 | - | 1 - 2 | 59 | 70 | ||
AK165420.1 | - | 1 - 2 | 69 | 70 | ||
AK165994.1 | - | 1 - 2 | 44 | 70 | ||
BB660164.1 | - | 1 - 2 | 114 | 70 | ||
BC026148.1 | - | 1 - 2 | 94 | 3492937 | 70 | |
BC033282.1 | - | 1 - 2 | 45 | 70 | ||
U51907.1 | - | 1 - 2 | 51 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK088817.1, AK165420.1, BC026148.1, NM_001164072.1, U51907.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
