Assay Details
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164072.1 | NP_001157544.1 | 2 - 3 | 263 | 81 | |
GenBank mRNA | AK088817.1 | - | 2 - 3 | 209 | 81 | |
AK165420.1 | - | 2 - 3 | 219 | 81 | ||
AK165994.1 | - | 2 - 3 | 194 | 81 | ||
BB660164.1 | - | 2 - 3 | 264 | 81 | ||
BC026148.1 | - | 2 - 3 | 244 | 3492937 | 81 | |
U51907.1 | - | 2 - 3 | 201 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
