Assay Details
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011112.3 | NP_035242.1 | 20 - 21 | 2249 | 79 | |
GenBank mRNA | AK028940.1 | - | 20 - 21 | 2247 | 79 | |
AK090096.1 | - | 20 - 21 | 2250 | 79 | ||
AK132704.1 | - | 20 - 21 | 2236 | 79 | ||
BC060128.1 | - | 19 - 20 | 2096 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
