Assay Details
Gene Symbol: Arhgap15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040796.1 | - | 5 - 6 | 451 | 80 | |
AK172407.1 | - | 5 - 6 | 450 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
