Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200007877.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007877.mm.1 |
NM_001043335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007877.mm.1 |
NM_001043335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007876.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007876.mm.1 |
NM_001043335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007876.mm.1 |
NM_001043335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007880.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007880.mm.1 |
NM_001043335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007880.mm.1 |
NM_001043335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007880.mm.1 |
NM_001043336.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007880.mm.1 |
NM_001286346.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007880.mm.1 |
NM_001286347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007875.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007875.mm.1 |
NM_001043335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007875.mm.1 |
NM_001043335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007879.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007879.mm.1 |
NM_001043335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007879.mm.1 |
NM_001043335.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007879.mm.1 |
NM_001043336.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007879.mm.1 |
NM_001286346.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007879.mm.1 |
NM_001286347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007873.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007873.mm.1 |
NM_001043336.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007873.mm.1 |
NM_001286347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007874.mm.1 |
TC1200001109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007874.mm.1 |
NM_001043335.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007874.mm.1 |
NM_001043335.1 |
Refseq |