Assay Details
Gene Symbol: 9030622O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018565.1 | - | 5 - 6 | 866 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA A530006G24 gene | A530006G24Rik | Gm1003 | -229.5kb | Anti-sense | Chr. 2 - 147710693 - 147717632 on NCBI Build GRCm38 |
Downstream | forkhead box A2 | Foxa2 |
HNF3-beta, HNF3beta, Hnf-3b, Hnf3b, Tcf-3b, Tcf3b |
80.8kb | Sense | Chr. 2 - 148042877 - 148046969 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
