Assay Details
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114088.2 | NP_001107560.1 | 8 - 9 | 725 | 95 | |
GenBank mRNA | AK033091.1 | - | 717 | 95 | ||
AK128987.1 | - | 7 - 8 | 1677 | 95 | ||
AK142933.1 | - | 8 - 9 | 709 | 97 | ||
AK167476.1 | - | 8 - 9 | 717 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
