Assay Details
Gene Symbol: Pik3r6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036623.1 | - | 2 - 3 | 260 | 66 | |
AK151178.1 | - | 2 - 3 | 244 | 66 | ||
AK153039.1 | - | 2 - 3 | 244 | 66 | ||
AK169948.1 | - | 2 - 3 | 262 | 66 | ||
AK171063.1 | - | 2 - 3 | 246 | 66 | ||
BC028998.1 | - | 2 - 3 | 252 | 5356085 | 66 | |
BY202586.1 | - | 2 - 3 | 324 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK151178.1, AK153039.1, AK171063.1, BC028998.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
