Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080853.3 | NP_543129.3 | 10 - 11 | 2090 | 112 | |
GenBank mRNA | AF324864.1 | - | 10 - 11 | 1923 | 112 | |
BC038375.1 | - | 10 - 11 | 1469 | 5357143 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
