Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080853.3 | NP_543129.3 | 5 - 6 | 1468 | 108 | |
GenBank mRNA | AF324864.1 | - | 5 - 6 | 1301 | 108 | |
AK045409.1 | - | 5 - 6 | 1231 | 108 | ||
BC038375.1 | - | 5 - 6 | 847 | 5357143 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
