Assay Details
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 13 - 14 | 1894 | 111 | |
GenBank mRNA | AK029009.1 | - | 2 - 3 | 96 | 111 | |
AK030158.1 | - | 5 - 6 | 593 | 111 | ||
L11625.1 | - | 4 - 5 | 432 | 111 | ||
U21301.1 | - | 13 - 14 | 1894 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
