Assay Details
Gene Symbol: Pnpla7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146251.4 | NP_666363.3 | 21 - 22 | 2590 | 77 | |
GenBank mRNA | AK038505.1 | - | 21 - 22 | 2476 | 77 | |
AK088362.1 | - | 21 - 22 | 2536 | 77 | ||
AK148380.1 | - | 21 - 22 | 2564 | 77 | ||
AK170417.1 | - | 21 - 22 | 2536 | 77 | ||
BC021312.1 | - | 20 - 21 | 2624 | 77 | ||
BC025621.1 | - | 17 - 18 | 2003 | 77 | ||
BC064003.1 | - | 4 - 5 | 354 | 77 | ||
BC141365.1 | - | 21 - 22 | 2374 | 9055978 | 77 | |
BC141366.1 | - | 21 - 22 | 2374 | 9055979 | 77 | |
EU293208.1 | - | 20 - 21 | 2261 | 77 | ||
EU368675.1 | - | 20 - 21 | 2261 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
