Assay Details
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016719.1 | NP_057928.1 | 10 - 11 | 1332 | 67 | |
GenBank mRNA | AF155647.1 | - | 10 - 11 | 1332 | 67 | |
AK147041.1 | - | 10 - 11 | 1341 | 67 | ||
BC021820.1 | - | 3 - 4 | 218 | 67 | ||
BC138581.1 | - | 10 - 11 | 1238 | 8861601 | 67 | |
BC145913.1 | - | 10 - 11 | 1238 | 40131216 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
