Assay Details
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207206.2 | NP_997089.1 | 13 - 14 | 1501 | 64 | |
GenBank mRNA | BC046276.1 | - | 11 - 12 | 1388 | 64 | |
BC068131.1 | - | 13 - 14 | 1501 | 6414152 | 64 | |
BC080829.1 | - | 13 - 14 | 1468 | 30435722 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
phospholipid metabolic process
metabolic process
phospholipid biosynthetic process
transferase activity
acyltransferase activity
1-alkenylglycerophosphoethanolamine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
lysophospholipid acyltransferase activity
lipid metabolic process
phospholipid metabolic process
metabolic process
phospholipid biosynthetic process
transferase activity
acyltransferase activity
1-alkenylglycerophosphoethanolamine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
lysophospholipid acyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527758 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527759 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527757 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527762 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527760 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087958 | BC046276.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087958 | BC068131.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087958 | BC080829.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087958 | NM_207206.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343398 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600465 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95576 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147756 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087958 | BC046276.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087958 | BC068131.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087958 | BC080829.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087958 | NM_207206.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527774 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21527766 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147756 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04647923 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134266 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102766 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147756 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01369118 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02243589 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01369115 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01369119 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527757 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527758 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527759 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527760 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21527762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01369115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02215860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02088490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01848496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01474492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01474494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01296672 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02213564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01474489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02271344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02243581 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01569456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02243585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01474482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02243589 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01259375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02215863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01569458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527823 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21527755 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10474537 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474529 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474531 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474527 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474533 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474528 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474535 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474538 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474534 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474530 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474532 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10474536 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440207_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10474526 | - | - |
Mouse Genome 430 2.0 Array | 1440207_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440207_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 99010_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 99010_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200012336.mm.1 | TC0200001662.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200012336.mm.1 | NM_207206.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200012336.mm.1 | NM_207206.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200012337.mm.1 | TC0200001662.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200012337.mm.1 | NM_207206.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200012337.mm.1 | NM_207206.2 | Refseq |
Mouse Expression Set 430 | 1440207_PM_at | - | - |
Mouse Expression Set 430 | 1440207_at | - | - |
Murine Genome U74v2 Set | 135603_at | - | - |
Murine Genome U74v2 Set | 135603_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440207_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440207_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 658007 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2810603 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002810603 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756891 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2756891 | - | - |
mouse-6 v1.1 (Illumina) | scl20489.13.1_48-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2810603 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20489.13.1_48-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810603 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2756891 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2756891 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20489.13.1_48-S | - | - |
