Assay Details
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207206.2 | NP_997089.1 | 6 - 7 | 813 | 88 | |
GenBank mRNA | BC046276.1 | - | 4 - 5 | 700 | 88 | |
BC068131.1 | - | 6 - 7 | 813 | 6414152 | 88 | |
BC080829.1 | - | 6 - 7 | 780 | 30435722 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
