Assay Details
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207206.2 | NP_997089.1 | 9 - 10 | 991 | 63 | |
GenBank mRNA | BC046276.1 | - | 7 - 8 | 878 | 63 | |
BC068131.1 | - | 9 - 10 | 991 | 6414152 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
