Assay Details
Gene Symbol: Dennd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146122.3 | NP_666234.3 | 7 - 8 | 705 | 61 | |
GenBank mRNA | AK137530.1 | - | 6 - 7 | 633 | 61 | |
AK220407.1 | - | 5 - 6 | 453 | 61 | ||
BC023016.1 | - | 4 - 5 | 447 | 61 | ||
BQ444119.1 | - | 7 - 8 | 543 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
