Assay Details
Gene Symbol: Hc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010406.2 | NP_034536.1 | 4 - 5 | 552 | 138 | |
GenBank mRNA | AK080166.1 | - | 4 - 5 | 551 | 138 | |
M35525.1 | - | 4 - 5 | 507 | 138 | ||
M35526.1 | - | 4 - 5 | 507 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
