Assay Details
Gene Symbol: Rps6ka5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030051.1 | - | 11 - 12 | 1561 | 83 | |
AK032316.1 | - | 12 - 13 | 1713 | 83 | ||
AK046319.1 | - | 12 - 13 | 1397 | 83 | ||
AK160758.1 | - | 5 - 6 | 579 | 83 | ||
AY341881.1 | - | 12 - 13 | 1609 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
