Assay Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029617.2 | NP_083893.2 | 13 - 14 | 5433 | 141 | |
GenBank mRNA | AK017750.1 | - | 1 - 2 | 82 | 141 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK017750.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
