Assay Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029617.2 | NP_083893.2 | 5 - 6 | 324 | 103 | |
GenBank mRNA | AK009772.1 | - | 5 - 6 | 326 | 103 | |
AK033132.1 | - | 5 - 6 | 628 | 103 | ||
BC080815.1 | - | 5 - 6 | 642 | 30104270 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
