Assay Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029617.2 | NP_083893.2 | 7 - 8 | 426 | 68 | |
GenBank mRNA | AK009772.1 | - | 7 - 8 | 428 | 68 | |
AK033132.1 | - | 7 - 8 | 730 | 68 | ||
BC080815.1 | - | 7 - 8 | 744 | 30104270 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
