Assay Details
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022317.3 | NP_071712.3 | 16 - 17 | 1948 | 78 | |
GenBank mRNA | AF305211.1 | - | 16 - 17 | 1881 | 78 | |
AK052784.1 | - | 16 - 17 | 1949 | 78 | ||
AK145077.1 | - | 16 - 17 | 1949 | 78 | ||
BC010472.1 | - | 16 - 17 | 1936 | 3586005 | 78 | |
BC013783.1 | - | 16 - 17 | 1939 | 3600786 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
