Assay Details
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290424.1 | NP_001277353.1 | 13 - 14 | 1547 | 97 | |
NM_008031.3 | NP_032057.2 | 13 - 14 | 1547 | 97 | ||
GenBank mRNA | AF461114.1 | - | 13 - 14 | 1271 | 97 | |
AK140343.1 | - | 13 - 14 | 1509 | 97 | ||
BC079671.1 | - | 13 - 14 | 1533 | 30532682 | 97 | |
L23971.1 | - | 13 - 14 | 1394 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
