Assay Details
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015792.1 | NP_056607.1 | 19 - 20 | 2918 | 68 | |
GenBank mRNA | AF184275.1 | - | 17 - 18 | 2412 | 68 | |
AK003964.1 | - | 4 - 5 | 479 | 68 | ||
AK154357.1 | - | 19 - 20 | 2941 | 68 | ||
AK158937.1 | - | 19 - 20 | 2969 | 68 | ||
BC004036.1 | - | 16 - 17 | 2004 | 68 | ||
BC031393.1 | - | 19 - 20 | 2918 | 4948804 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
