Assay Details
Gene Symbol: Unc13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029873.2 | NP_001025044.2 | 4 - 5 | 383 | 92 | |
GenBank mRNA | AK010728.1 | - | 3 - 4 | 319 | 92 | |
AK170263.1 | - | 3 - 4 | 1663 | 92 | ||
AY753536.1 | - | 4 - 5 | 273 | 92 | ||
CJ106055.1 | - | 4 - 5 | 383 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
