Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290821.1 | NP_001277750.1 | 3 - 4 | 710 | 72 | |
NM_020492.4 | NP_065238.2 | 3 - 4 | 710 | 72 | ||
GenBank mRNA | AK078258.1 | - | 3 - 4 | 710 | 72 | |
AY129229.1 | - | 3 - 4 | 257 | 72 | ||
BC131949.1 | - | 3 - 4 | 375 | 40130226 | 72 | |
BC137725.1 | - | 3 - 4 | 375 | 8860744 | 72 | |
S73717.1 | - | 3 - 4 | 302 | 72 | ||
S73718.1 | - | 3 - 4 | 302 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
