Assay Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290821.1 | NP_001277750.1 | 7 - 8 | 1377 | 84 | |
NM_020492.4 | NP_065238.2 | 7 - 8 | 1377 | 84 | ||
GenBank mRNA | AK078258.1 | - | 7 - 8 | 1375 | 84 | |
AY129229.1 | - | 7 - 8 | 924 | 84 | ||
BC131949.1 | - | 7 - 8 | 1042 | 40130226 | 84 | |
BC137725.1 | - | 7 - 8 | 1042 | 8860744 | 84 | |
S73717.1 | - | 7 - 8 | 969 | 84 | ||
S73718.1 | - | 7 - 8 | 969 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
