Assay Details
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043344.1 | - | 13 - 14 | 1488 | 68 | |
AK145200.1 | - | 14 - 15 | 1544 | 68 | ||
AK161473.1 | - | 14 - 15 | 1627 | 68 | ||
AK166305.1 | - | 10 - 11 | 1053 | 68 | ||
BC030328.1 | - | 14 - 15 | 1544 | 5376020 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
