Assay Details
Gene Symbol: AI314180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172381.2 | NP_759013.2 | 23 - 24 | 2881 | 95 | |
GenBank mRNA | AK035657.1 | - | 2 - 3 | 240 | 95 | |
AK043020.1 | - | 24 - 25 | 2798 | 95 | ||
AK129127.1 | - | 6 - 7 | 821 | 95 | ||
BC058684.1 | - | 23 - 24 | 2881 | 6834912 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
