Assay Details
Gene Symbol: AI314180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172381.2 | NP_759013.2 | 33 - 34 | 3910 | 68 | |
GenBank mRNA | AK043020.1 | - | 34 - 35 | 3827 | 68 | |
AK129127.1 | - | 16 - 17 | 1850 | 68 | ||
BC025035.1 | - | 3 - 4 | 182 | 68 | ||
BC058684.1 | - | 33 - 34 | 3910 | 6834912 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC025035.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
