Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033351.2 | NP_001028523.1 | 2 - 3 | 1997 | 93 | |
NM_001276355.1 | NP_001263284.1 | 2 - 3 | 1997 | 93 | ||
GenBank mRNA | AK044171.1 | - | 2 - 3 | 1974 | 93 | |
AK138366.1 | - | 2 - 3 | 1998 | 93 | ||
AK173314.1 | - | 2 - 3 | 998 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
