Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033351.2 | NP_001028523.1 | 5 - 6 | 3304 | 110 | |
NM_001276355.1 | NP_001263284.1 | 5 - 6 | 3304 | 110 | ||
GenBank mRNA | AK138366.1 | - | 5 - 6 | 3305 | 110 | |
AK173314.1 | - | 5 - 6 | 2305 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
