Assay Details
Gene Symbol: Scn10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205321.1 | NP_001192250.1 | 18 - 19 | 3303 | 92 | |
NM_009134.3 | NP_033160.2 | 18 - 19 | 3300 | 92 | ||
GenBank mRNA | AJ623269.1 | - | 2 - 3 | 408 | 92 | |
AJ623270.1 | - | 2 - 3 | 405 | 92 | ||
AK157866.1 | - | 18 - 19 | 3300 | 92 | ||
AY538273.1 | - | 17 - 18 | 3230 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
