Assay Details
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011049.4 | NP_035179.1 | 11 - 12 | 1451 | 80 | |
GenBank mRNA | AK051425.1 | - | 11 - 12 | 1452 | 80 | |
AK141083.1 | - | 5 - 6 | 81 | |||
AK143518.1 | - | 11 - 12 | 1479 | 80 | ||
AK145483.1 | - | 11 - 12 | 1481 | 80 | ||
AK166188.1 | - | 11 - 12 | 1660 | 80 | ||
BC011069.1 | - | 11 - 12 | 1194 | 4006745 | 80 | |
BC013663.1 | - | 11 - 12 | 1510 | 4208974 | 80 | |
X69025.1 | - | 11 - 12 | 1473 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
