Assay Details
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113325.2 | NP_001106796.1 | 8 - 9 | 1554 | 121 | |
NM_001252403.1 | NP_001239332.1 | 8 - 9 | 1181 | 121 | ||
NM_008165.4 | NP_032191.2 | 8 - 9 | 1554 | 121 | ||
GenBank mRNA | AB102773.1 | - | 8 - 9 | 1138 | 121 | |
AB102774.1 | - | 8 - 9 | 1138 | 121 | ||
AK049958.1 | - | 8 - 9 | 1364 | 121 | ||
AK135107.1 | - | 8 - 9 | 1182 | 121 | ||
BC056397.1 | - | 8 - 9 | 1216 | 6842391 | 121 | |
BC060702.1 | - | 8 - 9 | 1137 | 121 | ||
X57497.1 | - | 8 - 9 | 1401 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
