Assay Details
Gene Symbol: Zc3h7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145931.2 | NP_666043.2 | 16 - 17 | 2211 | 136 | |
NR_027502.1 | 16 - 17 | 2211 | 136 | |||
GenBank mRNA | AK028438.1 | - | 16 - 17 | 2139 | 136 | |
AK040507.1 | - | 16 - 17 | 2159 | 136 | ||
AK049834.1 | - | 16 - 17 | 2146 | 136 | ||
AK150952.1 | - | 16 - 17 | 2130 | 136 | ||
AK166876.1 | - | 17 - 18 | 2299 | 136 | ||
AK167758.1 | - | 16 - 17 | 2160 | 136 | ||
BC027330.1 | - | 16 - 17 | 2107 | 3674968 | 136 | |
BC029320.1 | - | 16 - 17 | 2109 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
