Assay Details
Gene Symbol: Mcm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008565.3 | NP_032591.3 | 2 - 3 | 449 | 84 | |
GenBank mRNA | AK011743.1 | - | 2 - 3 | 384 | 84 | |
AK036653.1 | - | 2 - 3 | 408 | 84 | ||
AK083618.1 | - | 2 - 3 | 363 | 84 | ||
AK088796.1 | - | 2 - 3 | 369 | 84 | ||
AK089999.1 | - | 2 - 3 | 353 | 84 | ||
AK135422.1 | - | 2 - 3 | 351 | 84 | ||
AK141132.1 | - | 2 - 3 | 415 | 84 | ||
AK141644.1 | - | 2 - 3 | 450 | 84 | ||
AK146662.1 | - | 2 - 3 | 372 | 84 | ||
AK151499.1 | - | 2 - 3 | 363 | 84 | ||
AK157725.1 | - | 2 - 3 | 351 | 84 | ||
AK220442.1 | - | 2 - 3 | 350 | 84 | ||
BC013094.1 | - | 2 - 3 | 405 | 3584640 | 84 | |
D26089.1 | - | 2 - 3 | 355 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
