Assay Details
Gene Symbol: Mcm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008565.3 | NP_032591.3 | 4 - 5 | 716 | 74 | |
GenBank mRNA | AK011743.1 | - | 4 - 5 | 651 | 74 | |
AK036653.1 | - | 4 - 5 | 676 | 74 | ||
AK083618.1 | - | 4 - 5 | 630 | 74 | ||
AK088796.1 | - | 4 - 5 | 636 | 74 | ||
AK089999.1 | - | 4 - 5 | 620 | 74 | ||
AK135422.1 | - | 4 - 5 | 618 | 74 | ||
AK141132.1 | - | 4 - 5 | 682 | 74 | ||
AK141644.1 | - | 4 - 5 | 717 | 74 | ||
AK146662.1 | - | 4 - 5 | 639 | 74 | ||
AK151499.1 | - | 4 - 5 | 630 | 74 | ||
AK157725.1 | - | 4 - 5 | 618 | 74 | ||
AK220442.1 | - | 4 - 5 | 617 | 74 | ||
BC013094.1 | - | 4 - 5 | 672 | 3584640 | 74 | |
D26089.1 | - | 4 - 5 | 622 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
