Assay Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF402611.1 | - | 16 - 17 | 2374 | 95 | |
AK028302.1 | - | 13 - 14 | 1823 | 95 | ||
AK149626.1 | - | 16 - 17 | 2425 | 95 | ||
BC129902.1 | - | 16 - 17 | 2461 | 40111637 | 95 | |
BC129903.1 | - | 16 - 17 | 2462 | 40111639 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



MicroArray Probes:
