Assay Details
Gene Symbol: Phldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252442.1 | NP_001239371.1 | 16 - 17 | 3645 | 91 | |
NM_153412.4 | NP_700461.2 | 15 - 16 | 3486 | 91 | ||
GenBank mRNA | AK035655.1 | - | 10 - 11 | 1249 | 91 | |
BC050915.1 | - | 16 - 17 | 3501 | 6405379 | 91 | |
BC060683.1 | - | 16 - 17 | 3501 | 6405379 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
