Assay Details
Gene Symbol: Cblb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033238.1 | NP_001028410.1 | 4 - 5 | 722 | 72 | |
GenBank mRNA | AK147367.1 | - | 4 - 5 | 707 | 72 | |
AK156295.1 | - | 5 - 6 | 753 | 72 | ||
AK161486.1 | - | 2 - 3 | 308 | 72 | ||
AK171350.1 | - | 4 - 5 | 667 | 72 | ||
BC150934.1 | - | 4 - 5 | 678 | 9087845 | 72 | |
BC150938.1 | - | 4 - 5 | 678 | 9087849 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
